Gene Transcripts

Gene Symbol
SDCCAG3
Entrez Gene ID
10807
Gene Name
serologically defined colon cancer antigen 3
Gene Aliases
NY-CO-3
Chromosome Location
Chr. 9: 136401922 - 136410602 on Build GRCh38
UniGene ID
Hs.94300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039707.1 NP_001034796.1 10 1548
NM_001039708.1 NP_001034797.1 8 1329
NM_006643.3 NP_006634.3 9 1479
XM_005266050.3 XP_005266107.1 10 1475
XM_005266051.3 XP_005266108.1 9 1393
XM_011518157.2 XP_011516459.1 9 1332
XM_017014218.1 XP_016869707.1 9 1649
XM_017014219.1 XP_016869708.1 9 1622
GenBank mRNA AF039688.1 - 6 616
AK001296.1 - 8 1108
AY129014.1 - 3 1422
BC014515.1 - 9 1346 3547130

View Full Product Details