Gene Transcripts

Gene Symbol
IRAK1
Entrez Gene ID
3654
Gene Name
interleukin 1 receptor associated kinase 1
Gene Aliases
IRAK, pelle
Chromosome Location
Chr. X: 154010506 - 154019906 on Build GRCh38
UniGene ID
Hs.522819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025242.1 NP_001020413.1 6, 7 881
NM_001025243.1 NP_001020414.1 6, 7 881
NM_001569.3 NP_001560.2 6, 7 881
XM_005274668.3 XP_005274725.1 5, 6 903
XM_011531158.2 XP_011529460.1 6, 7 896
GenBank mRNA AF346607.1 - 6, 7 802
BC014963.1 - 6, 7 866 4906853
BC054000.1 - 5, 6 958 6164719
DQ054788.1 - 6, 7 802
L76191.1 - 6, 7 881

View Full Product Details