Gene Transcripts

Gene Symbol
KPTN
Entrez Gene ID
11133
Gene Name
kaptin, actin binding protein
Gene Aliases
2E4, MRT41
Chromosome Location
Chr. 19: 47475141 - 47486792 on Build GRCh38
UniGene ID
Hs.25441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291296.1 NP_001278225.1 1 246
NM_007059.3 NP_008990.2 1 246
NR_111923.1 1 246
XM_011526399.2 XP_011524701.1 4 1751
XM_017026226.1 XP_016881715.1 4 596
XM_017026227.1 XP_016881716.1 3 672
XM_017026228.1 XP_016881717.1 4 596
XM_017026229.1 XP_016881718.1 4 596
GenBank mRNA AF105369.1 - 1 178
AK023996.1 - 1 201
AK225419.1 - 1 190
AK298672.1 - 1 144
BC009249.2 - 1 161 3029623
DA129265.1 - 1 246
DA824599.1 - 1 250
DR759729.1 - 1 158

View Full Product Details