Gene Transcripts

Gene Symbol
OR52A1
Entrez Gene ID
23538
Gene Name
olfactory receptor family 52 subfamily A member 1
Gene Aliases
HPFH1OR
Chromosome Location
Chr. 11: 5151431 - 5152369 on Build GRCh38
UniGene ID
Hs.553520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012375.2 NP_036507.2 1 431
GenBank mRNA BC111722.1 - 1 431

View Full Product Details