Gene Transcripts

Gene Symbol
COBL
Entrez Gene ID
23242
Gene Name
cordon-bleu WH2 repeat protein
Gene Aliases
Chromosome Location
Chr. 7: 51016212 - 51316818 on Build GRCh38
UniGene ID
Hs.99141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287436.1 NP_001274365.1 11, 12 3733
NM_015198.3 NP_056013.2 10, 11 3562
XM_005271750.1 XP_005271807.1 12, 13 3769
XM_005271751.1 XP_005271808.1 11, 12 3724
XM_005271756.1 XP_005271813.1 10, 11 3553
XM_011515234.1 XP_011513536.1 13, 14 3844
XM_011515235.1 XP_011513537.1 12, 13 3799
XM_011515236.1 XP_011513538.1 13, 14 3844
XM_011515237.1 XP_011513539.1 12, 13 3673
XM_011515238.1 XP_011513540.1 13, 14 3844
XM_011515239.1 XP_011513541.1 12, 13 3673
XM_011515240.1 XP_011513542.1 11, 12 3628
XM_011515241.1 XP_011513543.1 7, 8 2848
XM_017011898.1 XP_016867387.1 11, 12 3598
XM_017011899.1 XP_016867388.1 11, 12 3724
XM_017011900.1 XP_016867389.1 10, 11 3553
GenBank mRNA AB014533.1 - 10, 11 3543
AK308718.1 - 1, 2 693
AL713786.1 - 2, 3 7017
BC029275.1 - 5, 6 2433
BC045771.1 - 9, 10 3032
BC136441.1 - 11, 12 3648 9020430
BC144099.1 - 10, 11 3477 9052619
BC150263.1 - 10, 11 3543 9007207
CR936689.1 - 9, 10 3053

View Full Product Details