Gene Transcripts

Gene Symbol
POFUT2
Entrez Gene ID
23275
Gene Name
protein O-fucosyltransferase 2
Gene Aliases
C21orf80, FUT13
Chromosome Location
Chr. 21: 45263928 - 45287912 on Build GRCh38
UniGene ID
Hs.592164
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015227.4 NP_056042.1 3, 4 546
NM_133635.4 NP_598368.2 3, 4 546
NR_004858.1 3, 4 546
XM_006723981.3 XP_006724044.1 4, 5 619
GenBank mRNA AB023175.1 - 3, 4 535
AJ302079.2 - 3, 4 535
AJ302080.2 - 3, 4 535
AJ575591.1 - 3, 4 530
AK290732.1 - 3, 4 543
AK298080.1 - 3, 4 543
AL110285.1 - 3, 4 532
AY066015.2 - 3, 4 535
BC064623.1 - 3, 4 528 5727996

View Full Product Details