Gene Transcripts

Gene Symbol
NAPA
Entrez Gene ID
8775
Gene Name
NSF attachment protein alpha
Gene Aliases
SNAPA
Chromosome Location
Chr. 19: 47487634 - 47515258 on Build GRCh38
UniGene ID
Hs.126938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003827.3 NP_003818.2 2 447
NR_038456.1 2 447
XM_011527436.1 XP_011525738.1 2 121
GenBank mRNA AK098401.1 - 2 280
AK292244.1 - 2 211
AK308633.1 - 3 337
AK310185.1 - 2 204
AY927609.1 - 2 229
BC001165.1 - 2 219 3533668
BC007432.2 - 2 210 2962847
BC028234.1 - 2 265 5245770
BC091511.1 - 2 226 30512911
BQ948081.1 - 2 429
BT019568.1 - 2 129
U39412.1 - 2 196

View Full Product Details