Gene Transcripts
- Gene Symbol
- PEX11A
- Entrez Gene ID
- 8800
- Gene Name
- peroxisomal biogenesis factor 11 alpha
- Gene Aliases
- PEX11-ALPHA, PMP28, hsPEX11p
- Chromosome Location
- Chr. 15: 89681531 - 89690784 on Build GRCh38
- UniGene ID
- Hs.31034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003847.2 |
NP_003838.1
|
2, 3 |
318 |
|
GenBank mRNA |
AB015594.1 |
- |
2, 3 |
261 |
|
AF093668.1 |
- |
2, 3 |
223 |
|
AK001415.1 |
- |
2, 3 |
311 |
|
AK313318.1 |
- |
2, 3 |
232 |
|
BC009697.1 |
- |
2, 3 |
212 |
3878406 |
BC101164.2 |
- |
2, 3 |
172 |
40015908 |
BC114384.1 |
- |
2, 3 |
172 |
40004153 |
CR542046.1 |
- |
2, 3 |
166 |
|
CR542075.1 |
- |
2, 3 |
166 |
|
View Full Product Details