Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276336.1 | NP_001263265.1 | 4 | 501 | |
NM_001276337.1 | NP_001263266.1 | 3 | 366 | ||
NM_001282958.1 | NP_001269887.1 | 4 | 501 | ||
NM_004846.3 | NP_004837.1 | 4 | 501 | ||
XM_005246975.3 | XP_005247032.1 | 3 | 317 | ||
XM_011512206.2 | XP_011510508.1 | 4 | 776 | ||
XM_017005377.1 | XP_016860866.1 | 3 | 317 | ||
GenBank mRNA | AB209519.1 | - | 4 | 405 | |
AF038957.1 | - | 4 | 394 | ||
AF047695.1 | - | 4 | 392 | ||
AF068117.1 | - | 4 | 377 | ||
AK022518.1 | - | 4 | 399 | ||
AK303802.1 | - | 3 | 264 | ||
BC005392.1 | - | 4 | 399 | 3997986 | |
BC005874.2 | - | 4 | 421 | 2961437 | |
BC021226.2 | - | 4 | 365 | 4302323 | |
BC021690.1 | - | 4 | 403 | 3961350 | |
BG913475.1 | - | 4 | 402 | ||
CB994873.1 | - | 469 | |||
CD684451.1 | - | 3 | 289 | ||
DA408433.1 | - | 2 | 121 | ||
DA992544.1 | - | 4 | 501 |