Gene Transcripts

Gene Symbol
ZMYM4
Entrez Gene ID
9202
Gene Name
zinc finger MYM-type containing 4
Gene Aliases
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location
Chr. 1: 35268694 - 35421944 on Build GRCh38
UniGene ID
Hs.269211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005095.2 NP_005086.2 27 4130
XM_005271328.4 XP_005271385.1 27 4409
XM_005271331.2 XP_005271388.1 24 3375
XM_011542421.2 XP_011540723.1 27 4056
XM_011542423.2 XP_011540725.1 28 4461
XM_011542425.2 XP_011540727.1 25 3415
XM_017002803.1 XP_016858292.1 28 4183
XM_017002804.1 XP_016858293.1 28 4123
XM_017002805.1 XP_016858294.1 23 3621
GenBank mRNA AB007885.1 - 23 3250
AK309705.1 - 26 3790
BC012093.1 - 25 3709
BC127113.1 - 28 4136 40125110
BC127114.1 - 24 3319 40125115
BX647376.1 - 23 3361
BX648067.1 - 18 2424
CR936741.1 - 26 3994

View Full Product Details