Gene Transcripts
- Gene Symbol
- REC8
- Entrez Gene ID
- 9985
- Gene Name
- REC8 meiotic recombination protein
- Gene Aliases
- HR21spB, REC8L1, Rec8p
- Chromosome Location
- Chr. 14: 24171853 - 24180408 on Build GRCh38
- UniGene ID
- Hs.419259
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001048205.1 |
NP_001041670.1
|
8, 9 |
1227 |
|
NM_005132.2 |
NP_005123.2
|
9, 10 |
1126 |
|
XM_011537421.1 |
XP_011535723.1
|
8, 9 |
1399 |
|
XM_017021841.1 |
XP_016877330.1
|
8, 9 |
1196 |
|
GenBank mRNA |
AF006264.1 |
- |
8, 9 |
699 |
|
AK001435.1 |
- |
9, 10 |
1126 |
|
AK291191.1 |
- |
9, 10 |
1258 |
|
AL832973.1 |
- |
8, 9 |
1117 |
|
BC004159.2 |
- |
8, 9 |
783 |
2822680 |
BC010887.1 |
- |
8, 9 |
1160 |
3535425 |
BC019326.1 |
- |
2, 3 |
145 |
|
CR457263.1 |
- |
8, 9 |
699 |
|
View Full Product Details