Gene Transcripts
- Gene Symbol
- GPC6
- Entrez Gene ID
- 10082
- Gene Name
- glypican 6
- Gene Aliases
- OMIMD1
- Chromosome Location
- Chr. 13: 93226825 - 94408020 on Build GRCh38
- UniGene ID
- Hs.444329
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005708.3 |
NP_005699.1
|
2, 3 |
958 |
|
XM_011521044.2 |
XP_011519346.1
|
2, 3 |
283 |
|
XM_017020298.1 |
XP_016875787.1
|
2, 3 |
2566 |
|
XM_017020299.1 |
XP_016875788.1
|
2, 3 |
1333 |
|
XM_017020300.1 |
XP_016875789.1
|
2, 3 |
190 |
|
GenBank mRNA |
AF105267.1 |
- |
2, 3 |
911 |
|
AF111178.1 |
- |
2, 3 |
960 |
|
AK074741.1 |
- |
2, 3 |
412 |
|
AK130013.1 |
- |
2, 3 |
295 |
|
AK290144.1 |
- |
2, 3 |
479 |
|
AK304338.1 |
- |
2, 3 |
777 |
|
AY358462.1 |
- |
2, 3 |
891 |
|
BC106947.2 |
- |
2, 3 |
806 |
40034564 |
View Full Product Details