Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005777.2 | NP_005768.1 | 3 | 644 | |
XM_005264784.1 | XP_005264841.1 | 3 | 919 | ||
XM_005264787.2 | XP_005264844.1 | 1 | 73 | ||
XM_006712916.1 | XP_006712979.1 | 3 | 744 | ||
XM_017005496.1 | XP_016860985.1 | 1 | 73 | ||
XM_017005499.1 | XP_016860988.1 | 3 | 919 | ||
XM_017005500.1 | XP_016860989.1 | 3 | 919 | ||
XM_017005501.1 | XP_016860990.1 | 3 | 919 | ||
GenBank mRNA | AF042857.1 | - | 3 | 497 | |
AF069517.1 | - | 3 | 600 | ||
AF091264.1 | - | 3 | 529 | ||
AK291719.1 | - | 3 | 644 | ||
AK303371.1 | - | 3 | 537 | ||
BX648535.1 | - | 3 | 821 | ||
U50839.1 | - | 2 | 430 |