Gene Transcripts

Gene Symbol
TAAR2
Entrez Gene ID
9287
Gene Name
trace amine associated receptor 2
Gene Aliases
GPR58, taR-2
Chromosome Location
Chr. 6: 132617150 - 132624275 on Build GRCh38
UniGene ID
Hs.272382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033080.1 NP_001028252.1 2 640
NM_014626.3 NP_055441.2 1 528
GenBank mRNA AY702304.1 - 1 505
AY703480.1 - 2 640
BC067461.1 - 1 505
BC067462.1 - 1 505
BC067463.1 - 1 505
DN693314.1 - 1 528

View Full Product Details