Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199787.1 | NP_001186716.1 | 4 | 1217 | |
NM_182838.2 | NP_878258.1 | 3 | 799 | ||
GenBank mRNA | AL049665.1 | - | 3 | 792 | |
BC101666.1 | - | 2 | 398 | 8069172 | |
BC112289.1 | - | 2 | 398 | 8327757 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110781.2 | NP_001104251.1 | 3 | 879 | |
NM_001290264.1 | NP_001277193.1 | 4 | 1297 | ||
XM_011542072.1 | XP_011540374.1 | 2 | 488 | ||
GenBank mRNA | AB007916.3 | - | 3 | 817 | |
AK125619.1 | - | 3 | 803 | ||
AK307886.1 | - | 2 | 481 | ||
BC110653.1 | - | 3 | 855 | 4392812 | |
CR936618.1 | - | 4 | 1017 |