Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267580.1 | NP_001254509.1 | 2 | 341 | |
NM_003981.3 | NP_003972.1 | 2 | 341 | ||
NM_199413.2 | NP_955445.1 | 2 | 341 | ||
XM_005254987.2 | XP_005255044.1 | 2 | 249 | ||
XM_006720759.1 | XP_006720822.1 | 2 | 249 | ||
XM_006720760.1 | XP_006720823.1 | 2 | 249 | ||
XM_011522187.1 | XP_011520489.1 | 2 | 249 | ||
XM_011522188.2 | XP_011520490.1 | 2 | 249 | ||
XM_011522189.1 | XP_011520491.1 | 2 | 249 | ||
XM_011522190.2 | XP_011520492.1 | 2 | 249 | ||
XM_011522191.2 | XP_011520493.1 | 2 | 249 | ||
XM_011522192.1 | XP_011520494.1 | 2 | 249 | ||
XM_017022712.1 | XP_016878201.1 | 2 | 249 | ||
XM_017022713.1 | XP_016878202.1 | 2 | 249 | ||
XM_017022716.1 | XP_016878205.1 | 2 | 249 | ||
GenBank mRNA | AF044588.1 | - | 2 | 185 | |
AK297117.1 | - | 2 | 215 | ||
AK312600.1 | - | 2 | 240 | ||
BC003138.1 | - | 2 | 209 | 2958690 | |
BC005140.2 | - | 2 | 239 | 3504761 | |
BG773441.1 | - | 2 | 297 | ||
BX457401.2 | - | 2 | 194 |