Gene Transcripts

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
Chromosome Location
Chr. 1: 74198212 - 74208702 on Build GRCh38
UniGene ID
Hs.480085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199328.2 NP_001186257.2 2 222
NM_001199329.2 NP_001186258.2 2 222
NM_003838.4 NP_003829.3 2 222
GenBank mRNA AF017445.1 - 2 192
AK223254.1 - 2 189
AK294948.1 - 2 194
AK299468.1 - 2 167
AK304490.1 - 2 190
BC020720.1 - 2 196
BC032308.1 - 2 178 5184469
DB066016.1 - 2 226
KU178439.1 - 2 155
KU178440.1 - 2 155
KU178441.1 - 2 155

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
Chromosome Location
Chr. 1: 74198212 - 74544432 on Build GRCh38
UniGene ID
Hs.480085
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112808.2 NP_001106279.2 2 222
NM_001199327.1 NP_001186256.2 2 222
GenBank mRNA AB208941.1 - 2 171
BC032865.1 - 2 195 5263275
BX640903.1 - 2 207

View Full Product Details