Gene Transcripts

Gene Symbol
TRPC3
Entrez Gene ID
7222
Gene Name
transient receptor potential cation channel subfamily C member 3
Gene Aliases
SCA41, TRP3
Chromosome Location
Chr. 4: 121879028 - 121952060 on Build GRCh38
UniGene ID
Hs.150981
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130698.1 NP_001124170.1 10 2551
NM_003305.2 NP_003296.1 9 2333
XM_011532217.2 XP_011530519.1 11 2905
XM_011532218.2 XP_011530520.1 10 2857
XM_017008578.1 XP_016864067.1 11 2905
GenBank mRNA AB255423.1 - 8 2093
AB255424.1 - 9 2258
AY865574.1 - 10 2551
BC093682.1 - 9 2333 7939527
BC093684.1 - 9 2333 7939529
Y13758.1 - 9 2682

View Full Product Details