Gene Transcripts
- Gene Symbol
- PIP4K2B
- Entrez Gene ID
- 8396
- Gene Name
- phosphatidylinositol-5-phosphate 4-kinase type 2 beta
- Gene Aliases
- PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB
- Chromosome Location
- Chr. 17: 38765691 - 38799965 on Build GRCh38
- UniGene ID
- Hs.260603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003559.4 |
NP_003550.1
|
5, 6 |
1137 |
|
XM_011525326.2 |
XP_011523628.1
|
6, 7 |
1278 |
|
XM_011525327.2 |
XP_011523629.1
|
7, 8 |
797 |
|
XM_011525329.2 |
XP_011523631.1
|
6, 7 |
1278 |
|
XM_011525330.2 |
XP_011523632.1
|
4, 5 |
473 |
|
XM_017025196.1 |
XP_016880685.1
|
6, 7 |
714 |
|
XM_017025197.1 |
XP_016880686.1
|
6, 7 |
614 |
|
XM_017025198.1 |
XP_016880687.1
|
4, 5 |
1100 |
|
XM_017025199.1 |
XP_016880688.1
|
5, 6 |
552 |
|
GenBank mRNA |
BC027459.1 |
- |
5, 6 |
754 |
|
BT019614.1 |
- |
5, 6 |
656 |
|
U85245.1 |
- |
5, 6 |
1136 |
|
View Full Product Details