Gene Transcripts

Gene Symbol
SYMPK
Entrez Gene ID
8189
Gene Name
symplekin
Gene Aliases
SPK, SYM
Chromosome Location
Chr. 19: 45815442 - 45863290 on Build GRCh38
UniGene ID
Hs.515475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004819.2 NP_004810.2 9 1137
XM_005259286.1 XP_005259343.1 9 1090
XM_011527354.1 XP_011525656.1 10 1140
GenBank mRNA AB209024.1 - 9 3385
AB210010.1 - 9 982
AK300252.1 - 9 991
BC006536.2 - 8 844
BC006567.2 - 8 844
BC030214.1 - 9 979
BX649165.1 - 9 992
U49240.1 - 4 606
Y10931.1 - 9 955

View Full Product Details