Gene Transcripts
- Gene Symbol
- STAT2
- Entrez Gene ID
- 6773
- Gene Name
- signal transducer and activator of transcription 2
- Gene Aliases
- IMD44, ISGF-3, P113, STAT113
- Chromosome Location
- Chr. 12: 56341597 - 56360253 on Build GRCh38
- UniGene ID
- Hs.530595
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005419.3 |
NP_005410.1
|
9, 10 |
1154 |
|
NM_198332.1 |
NP_938146.1
|
9, 10 |
1142 |
|
XM_005269111.3 |
XP_005269168.2
|
9, 10 |
1062 |
|
XM_011538697.2 |
XP_011536999.1
|
9, 10 |
1062 |
|
XM_011538698.2 |
XP_011537000.1
|
9, 10 |
1023 |
|
XM_011538699.2 |
XP_011537001.1
|
9, 10 |
1062 |
|
XM_011538700.2 |
XP_011537002.1
|
3, 4 |
460 |
|
XM_017019904.1 |
XP_016875393.1
|
8, 9 |
1521 |
|
GenBank mRNA |
AK296939.1 |
- |
9, 10 |
1015 |
|
AK296940.1 |
- |
9, 10 |
1015 |
|
BC006092.1 |
- |
8, 9 |
941 |
|
BC051284.1 |
- |
8, 9 |
974 |
6460486 |
M97934.1 |
- |
9, 10 |
988 |
|
View Full Product Details