Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_036632.1 | 10 | 1815 | ||
GenBank mRNA | AF217995.1 | - | 10 | 1765 | |
AK124885.1 | - | 3 | 4375 | ||
AK131330.1 | - | 6 | 4792 | ||
AL137720.1 | - | 3 | 866 | ||
BC109389.1 | - | 6 | 850 | ||
BC110077.1 | - | 6 | 850 | ||
BC127181.1 | - | 6 | 920 | ||
BC141930.1 | - | 6 | 850 | 40029611 | |
BX647364.1 | - | 7 | 2549 |