Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001076785.2 | NP_001070253.1 | 12 | 4518 | |
NM_003983.5 | NP_003974.3 | 11 | 4431 | ||
XM_011523433.1 | XP_011521735.1 | 11 | 4533 | ||
XM_011523434.1 | XP_011521736.1 | 11 | 4598 | ||
XM_011523438.1 | XP_011521740.1 | 10 | 4988 | ||
XM_011523439.1 | XP_011521741.1 | 12 | 5230 | ||
XM_017023851.1 | XP_016879340.1 | 10 | 4558 | ||
GenBank mRNA | CR749291.1 | - | 11 | 4417 | |
CR749475.1 | - | 11 | 3241 | ||
D87432.1 | - | 11 | 4466 |