Gene Transcripts

Gene Symbol
C15orf41
Entrez Gene ID
84529
Gene Name
chromosome 15 open reading frame 41
Gene Aliases
HH114
Chromosome Location
Chr. 15: 36579603 - 36810260 on Build GRCh38
UniGene ID
Hs.48348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130010.2 NP_001123482.1 11 993
NM_001290232.1 NP_001277161.1 11 684
NM_001321756.1 NP_001308685.1 10 947
NM_001321757.1 NP_001308686.1 12 1027
NM_001321758.1 NP_001308687.1 9 882
NM_001321759.1 NP_001308688.1 11 993
NM_001321760.1 NP_001308689.1 10 887
NM_001321761.1 NP_001308690.1 11 1011
NM_032499.5 NP_115888.1 12 739
XM_017022676.1 XP_016878165.1 8 787
XM_017022677.1 XP_016878166.1 8 787
GenBank mRNA AF114263.1 - 12 714
AK315446.1 - 11 653
BC006254.2 - 11 752 3952242
BC042750.1 - 3 5098
BX647650.1 - 5 363

View Full Product Details