Gene Transcripts

Gene Symbol
MIR4724
Entrez Gene ID
100616248
Gene Name
microRNA 4724
Gene Aliases
mir-4724
Chromosome Location
Chr. 17: 31534883 - 31534971 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_039877.1 1 36

Gene Symbol
RAB11FIP4
Entrez Gene ID
84440
Gene Name
RAB11 family interacting protein 4
Gene Aliases
RAB11-FIP4
Chromosome Location
Chr. 17: 31391624 - 31538217 on Build GRCh38
UniGene ID
Hs.406788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303542.2 NP_001290471.2 13 5036
NM_032932.5 NP_116321.2 15 5329
GenBank mRNA AB058724.1 - 15 5018
AJ314646.2 - 15 5018
AK055019.1 - 1 1927
AY169244.1 - 15 5329

View Full Product Details