Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001292048.1 | NP_001278977.1 | 2 | 670 | |
NM_004162.4 | NP_004153.2 | 2 | 670 | ||
GenBank mRNA | AF464088.1 | - | 2 | 511 | |
AF498936.1 | - | 1 | 135 | ||
AK295992.1 | - | 2 | 345 | ||
AK309287.1 | - | 2 | 351 | ||
AK312618.1 | - | 2 | 384 | ||
BC001267.1 | - | 2 | 300 | 3463669 | |
BC018288.1 | - | 2 | 353 | 3454268 | |
CR536492.1 | - | 1 | 135 | ||
DA972638.1 | - | 2 | 459 | ||
M28215.1 | - | 1 | 191 | ||
X13903.1 | - | 1 | 135 |