Gene Transcripts
- Gene Symbol
- IL34
- Entrez Gene ID
- 146433
- Gene Name
- interleukin 34
- Gene Aliases
- C16orf77, IL-34
- Chromosome Location
- Chr. 16: 70579895 - 70660682 on Build GRCh38
- UniGene ID
- Hs.461214
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172771.1 |
NP_001166242.1
|
3, 4 |
709 |
|
NM_001172772.1 |
NP_001166243.1
|
3, 4 |
692 |
|
NM_152456.2 |
NP_689669.2
|
3, 4 |
709 |
|
XM_011522902.2 |
XP_011521204.1
|
3, 4 |
793 |
|
XM_011522903.2 |
XP_011521205.1
|
3, 4 |
575 |
|
GenBank mRNA |
AK172744.1 |
- |
2, 3 |
224 |
|
AK307727.1 |
- |
2, 3 |
344 |
|
AK314913.1 |
- |
2, 3 |
229 |
|
BC029804.1 |
- |
2, 3 |
537 |
5192904 |
CD248375.1 |
- |
3, 4 |
610 |
|
CD515627.1 |
- |
3, 4 |
714 |
|
EU599219.1 |
- |
2, 3 |
154 |
|
EU636994.1 |
- |
2, 3 |
154 |
|
View Full Product Details