Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172771.1 | NP_001166242.1 | 7 | 1108 | |
NM_001172772.1 | NP_001166243.1 | 7 | 1094 | ||
NM_152456.2 | NP_689669.2 | 7 | 1111 | ||
XM_011522902.2 | XP_011521204.1 | 7 | 1195 | ||
XM_011522903.2 | XP_011521205.1 | 7 | 977 | ||
GenBank mRNA | AK172744.1 | - | 6 | 626 | |
AK307727.1 | - | 6 | 746 | ||
AK314913.1 | - | 6 | 631 | ||
BC029804.1 | - | 6 | 939 | 5192904 | |
EU599219.1 | - | 6 | 553 | ||
EU636994.1 | - | 6 | 556 |