Gene Transcripts
- Gene Symbol
- GAS2L3
- Entrez Gene ID
- 283431
- Gene Name
- growth arrest specific 2 like 3
- Gene Aliases
- G2L3
- Chromosome Location
- Chr. 12: 100573661 - 100628288 on Build GRCh38
- UniGene ID
- Hs.20575
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001303130.1 |
NP_001290059.1
|
7, 8 |
799 |
|
NM_001303131.1 |
NP_001290060.1
|
8, 9 |
1286 |
|
NM_174942.2 |
NP_777602.1
|
8, 9 |
920 |
|
XM_005268804.3 |
XP_005268861.1
|
9, 10 |
989 |
|
XM_011538219.2 |
XP_011536521.1
|
8, 9 |
845 |
|
XM_011538220.2 |
XP_011536522.1
|
7, 8 |
2180 |
|
XM_011538221.2 |
XP_011536523.1
|
7, 8 |
1030 |
|
XM_017019206.1 |
XP_016874695.1
|
9, 10 |
1063 |
|
GenBank mRNA |
AK095594.1 |
- |
8, 9 |
1286 |
|
AK315189.1 |
- |
7, 8 |
777 |
|
BC043366.1 |
- |
8, 9 |
870 |
5104644 |
View Full Product Details