Gene Transcripts

Gene Symbol
PSMD13
Entrez Gene ID
5719
Gene Name
proteasome 26S subunit, non-ATPase 13
Gene Aliases
HSPC027, Rpn9, S11, p40.5
Chromosome Location
Chr. 11: 236675 - 252984 on Build GRCh38
UniGene ID
Hs.134688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002817.3 NP_002808.3 9, 10 1024
NM_175932.2 NP_787128.2 7, 8 1030
XM_011520235.2 XP_011518537.1 9, 10 1157
GenBank mRNA AB009398.1 - 9, 10 851
AF083245.1 - 9, 10 836
AF086708.1 - 9, 10 782
AF107836.1 - 2, 3 224
AF107837.1 - 9, 10 815
AK092845.1 - 1 27
AK094775.1 - 7, 8 861
AK222895.1 - 9, 10 855
AK295944.1 - 8, 9 821
AK314733.1 - 9, 10 1286
BC001100.2 - 9, 10 837 3506530
BC001747.1 - 9, 10 805 3535807
BT007307.1 - 9, 10 782
BX647314.1 - 8, 9 759

View Full Product Details