Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323281.1 | NP_001310210.1 | 2 | 516 | |
NM_001323282.1 | NP_001310211.1 | 2 | 516 | ||
NM_002895.4 | NP_002886.2 | 2 | 516 | ||
NM_183404.3 | NP_899662.1 | 2 | 516 | ||
XM_006723842.2 | XP_006723905.1 | 2 | 292 | ||
XM_011528955.2 | XP_011527257.1 | 2 | 292 | ||
XM_011528958.2 | XP_011527260.1 | 2 | 292 | ||
XM_017027992.1 | XP_016883481.1 | 2 | 330 | ||
GenBank mRNA | AK290380.1 | - | 2 | 292 | |
AK308096.1 | - | 2 | 276 | ||
AK310870.1 | - | 2 | 301 | ||
BC032247.1 | - | 2 | 299 | 4521266 | |
L14812.1 | - | 2 | 297 |