Gene Transcripts

Gene Symbol
SDHD
Entrez Gene ID
6392
Gene Name
succinate dehydrogenase complex subunit D
Gene Aliases
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS
Chromosome Location
Chr. 11: 112086824 - 112095801 on Build GRCh38
UniGene ID
Hs.356270
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276503.1 NP_001263432.1 3 326
NM_001276504.1 NP_001263433.1 3 354
NM_001276506.1 NP_001263435.1 5 601
NM_003002.3 NP_002993.1 4 471
NR_077060.1 5 525
GenBank mRNA AB006202.1 - 4 398
AK075360.1 - 4 397
BC005263.1 - 4 415 3930691
BC009574.1 - 4 396 4063808
BC012603.1 - 4 400 4108104
BC015188.1 - 4 422 4279214
BC015992.1 - 4 418 4292983
BC022350.1 - 4 410 4275516
BC070307.1 - 4 396 4277540
BC071755.1 - 4 414 4774525
BC071756.1 - 4 396 6450992
BF211358.1 - 3 279
BQ932668.1 - 4 479
BT007238.1 - 4 387
CD105572.1 - 3 266
CR456932.1 - 4 387
KU178325.1 - 4 387
KU178326.1 - 3 242

View Full Product Details