Gene Transcripts

Gene Symbol
PRCC
Entrez Gene ID
5546
Gene Name
papillary renal cell carcinoma (translocation-associated)
Gene Aliases
RCCP1, TPRC
Chromosome Location
Chr. 1: 156767482 - 156800819 on Build GRCh38
UniGene ID
Hs.516948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005973.4 NP_005964.3 7 1970
XM_005245313.2 XP_005245370.1 8 2066
XM_005245314.1 XP_005245371.1 6 1874
GenBank mRNA AK126403.1 - 2 1821
BC004913.2 - 7 1886 3535413
BC010450.1 - 7 1898 4179105
BU942864.1 - 4 576
X97124.1 - 7 1870

View Full Product Details