Gene Transcripts

Gene Symbol
PSKH1
Entrez Gene ID
5681
Gene Name
protein serine kinase H1
Gene Aliases
Chromosome Location
Chr. 16: 67893272 - 67929678 on Build GRCh38
UniGene ID
Hs.513683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006742.2 NP_006733.1 2 455
GenBank mRNA AJ272212.1 - 2 415
AK309272.1 - 2 455
BC062616.1 - 2 426 5188431
DA429976.1 - 2 455

View Full Product Details