Gene Transcripts

Gene Symbol
SPP2
Entrez Gene ID
6694
Gene Name
secreted phosphoprotein 2
Gene Aliases
SPP-24, SPP24
Chromosome Location
Chr. 2: 234050671 - 234077134 on Build GRCh38
UniGene ID
Hs.444488
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006944.2 NP_008875.1 7 656
XM_005246102.3 XP_005246159.1 7 687
XM_011511698.2 XP_011510000.1 7 687
XM_011511699.2 XP_011510001.1 7 687
XM_011511700.2 XP_011510002.1 7 687
GenBank mRNA AJ308099.1 - 7 656
BC069401.1 - 7 571 7262158
BC106705.1 - 7 571
BC112438.1 - 7 571
BC113972.1 - 6 446
U20530.1 - 7 571

View Full Product Details