Gene Transcripts

Gene Symbol
PGR
Entrez Gene ID
5241
Gene Name
progesterone receptor
Gene Aliases
NR3C3, PR
Chromosome Location
Chr. 11: 101029624 - 101130681 on Build GRCh38
UniGene ID
Hs.32405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000926.4 NP_000917.3 5 3089
NM_001202474.3 NP_001189403.1 5 2339
NM_001271161.2 NP_001258090.1 4 2033
NM_001271162.1 NP_001258091.1 5 1009
NR_073141.2 5 2339
NR_073142.2 4 2222
NR_073143.2 4 2033
XM_006718858.3 XP_006718921.1 5 3939
GenBank mRNA AB084248.1 - 4 2040
AB085683.1 - 4 2040
AB085843.1 - 4 2229
AB085844.1 - 4 2040
AB085845.1 - 5 2346
AF016381.1 - 5 2521
AK304853.1 - 5 1009
AY212933.1 - 2 1719
AY382151.1 - 4 705
M15716.1 - 5 2521
X51730.1 - 5 3800

View Full Product Details