Gene Transcripts

Gene Symbol
NEK2
Entrez Gene ID
4751
Gene Name
NIMA related kinase 2
Gene Aliases
HsPK21, NEK2A, NLK1, PPP1R111, RP67
Chromosome Location
Chr. 1: 211658256 - 211675630 on Build GRCh38
UniGene ID
Hs.153704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204182.1 NP_001191111.1 3 534
NM_001204183.1 NP_001191112.1 3 534
NM_002497.3 NP_002488.1 3 534
XM_005273147.1 XP_005273204.1 3 531
GenBank mRNA AK223353.1 - 3 495
AK225522.1 - 3 495
AK303092.1 - 4 397
AY045701.1 - 3 465
BC043502.1 - 3 522 5541026
BC052807.1 - 3 529
BC065932.1 - 3 524
BT019729.1 - 3 383
DC407297.1 - 3 534
U11050.1 - 3 517
Z25425.1 - 2 137
Z29066.1 - 3 465

View Full Product Details