Gene Transcripts
- Gene Symbol
- SNU13
- Entrez Gene ID
- 4809
- Gene Name
- SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5)
- Gene Aliases
- 15.5K, FA-1, FA1, NHP2L1, NHPX, OTK27, SNRNP15-5, SPAG12, SSFA1
- Chromosome Location
- Chr. 22: 41673933 - 41690492 on Build GRCh38
- UniGene ID
- Hs.182255
- Species
- Homo sapiens
- Gene Symbol
- ANXA2
- Entrez Gene ID
- 302
- Gene Name
- annexin A2
- Gene Aliases
- ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
- Chromosome Location
- Chr. 15: 60347151 - 60398025 on Build GRCh38
- UniGene ID
- Hs.511605
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309356.1 |
- |
|
112 |
|
View Full Product Details