Gene Transcripts
- Gene Symbol
- PCBP2
- Entrez Gene ID
- 5094
- Gene Name
- poly(rC) binding protein 2
- Gene Aliases
- HNRNPE2, HNRPE2, hnRNP-E2
- Chromosome Location
- Chr. 12: 53452102 - 53481162 on Build GRCh38
- UniGene ID
- Hs.546271
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128911.1 |
NP_001122383.1
|
9, 10 |
1042 |
|
NM_001128912.1 |
NP_001122384.1
|
9, 10 |
1030 |
|
NM_005016.5 |
NP_005007.2
|
9, 10 |
1042 |
|
NM_031989.4 |
NP_114366.1
|
9, 10 |
1030 |
|
GenBank mRNA |
AB208825.1 |
- |
8, 9 |
764 |
|
AK023529.1 |
- |
4, 5 |
1641 |
|
AK130583.1 |
- |
2, 3 |
184 |
|
AK292141.1 |
- |
9, 10 |
807 |
|
BC001155.1 |
- |
8, 9 |
768 |
3356341 |
BC035420.1 |
- |
9, 10 |
1015 |
|
BC071942.1 |
- |
9, 10 |
1032 |
6424433 |
X78136.1 |
- |
8, 9 |
714 |
|
View Full Product Details