Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324255.1 | NP_001311184.1 | 4 | 1514 | |
NM_005909.4 | NP_005900.2 | 5 | 1784 | ||
GenBank mRNA | AK096273.1 | - | 6 | 1607 | |
AK308147.1 | - | 5 | 1575 | ||
BC017240.2 | - | 5 | 1761 | ||
BC025240.1 | - | 5 | 1703 | ||
BC032516.1 | - | 6 | 1627 | ||
BC039822.1 | - | 5 | 1570 | ||
BC046114.1 | - | 5 | 1590 | ||
BC056145.1 | - | 5 | 1564 | ||
BC063669.1 | - | 5 | 1564 | ||
BC073993.1 | - | 5 | 1566 | ||
BC094834.1 | - | 5 | 1561 | ||
BC108733.1 | - | 5 | 1579 | ||
BC139918.1 | - | 5 | 1681 | ||
BC141853.1 | - | 4 | 1367 | ||
BC150196.1 | - | 5 | 1681 | ||
BX647077.1 | - | 5 | 1575 |