Gene Transcripts
- Gene Symbol
- MOCS1
- Entrez Gene ID
- 4337
- Gene Name
- molybdenum cofactor synthesis 1
- Gene Aliases
- MIG11, MOCOD, MOCODA
- Chromosome Location
- Chr. 6: 39904258 - 39934551 on Build GRCh38
- UniGene ID
- Hs.357128
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001075098.3 |
NP_001068566.1
|
7, 8 |
1007 |
|
NM_005943.5 |
NP_005934.2
|
6, 7 |
1011 |
|
NR_033233.1 |
|
6, 7 |
880 |
|
XM_011514632.2 |
XP_011512934.1
|
7, 8 |
918 |
|
XM_011514633.2 |
XP_011512935.1
|
7, 8 |
918 |
|
XM_011514634.2 |
XP_011512936.1
|
6, 7 |
815 |
|
XM_017010888.1 |
XP_016866377.1
|
6, 7 |
825 |
|
GenBank mRNA |
AF034374.1 |
- |
7, 8 |
890 |
|
AF214015.1 |
- |
1, 2 |
27 |
|
AF214023.1 |
- |
1, 2 |
27 |
|
AJ224328.1 |
- |
6, 7 |
1008 |
|
AK056740.1 |
- |
6, 7 |
826 |
|
AK091306.1 |
- |
6, 7 |
739 |
|
AK300300.1 |
- |
7, 8 |
880 |
|
BC036839.1 |
- |
7, 8 |
880 |
|
CX787499.1 |
- |
3, 4 |
288 |
|
View Full Product Details