Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142605.1 | NP_001136077.1 | 2 | 388 | |
NM_001258353.1 | NP_001245282.1 | 3 | 634 | ||
NM_001258354.1 | NP_001245283.1 | 3 | 497 | ||
NM_004247.3 | NP_004238.3 | 3 | 497 | ||
GenBank mRNA | AK126464.1 | - | 4 | 303 | |
AK128148.1 | - | 3 | 634 | ||
AK225335.1 | - | 3 | 257 | ||
AK293108.1 | - | 3 | 260 | ||
AK296367.1 | - | 2 | 191 | ||
AK297392.1 | - | 3 | 303 | ||
AK316098.1 | - | 2 | 208 | ||
BC002360.2 | - | 3 | 267 | 2960986 | |
CR456774.1 | - | 2 | 236 | ||
D21163.1 | - | 3 | 296 | ||
DC353274.1 | - | 3 | 497 |