Gene Transcripts
- Gene Symbol
- GAGE1
- Entrez Gene ID
- 2543
- Gene Name
- G antigen 1
- Gene Aliases
- CT4.1, GAGE-1
- Chromosome Location
- Chr. X: 49598992 - 49608536 on Build GRCh38
- UniGene ID
- Hs.278606
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102272.1 |
|
5 |
540 |
|
GenBank mRNA |
BC036094.1 |
- |
5 |
528 |
|
U19142.1 |
- |
5 |
462 |
|
View Full Product Details