Gene Transcripts
- Gene Symbol
- KPNA3
- Entrez Gene ID
- 3839
- Gene Name
- karyopherin subunit alpha 3
- Gene Aliases
- IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
- Chromosome Location
- Chr. 13: 49699307 - 49792921 on Build GRCh38
- UniGene ID
- Hs.527919
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002267.3 |
NP_002258.2
|
3, 4 |
625 |
|
XM_017020561.1 |
XP_016876050.1
|
3, 4 |
308 |
|
GenBank mRNA |
AF005263.1 |
- |
3, 4 |
222 |
|
AF034756.1 |
- |
3, 4 |
212 |
|
AK223480.1 |
- |
3, 4 |
247 |
|
AK290528.1 |
- |
3, 4 |
285 |
|
AK291000.1 |
- |
3, 4 |
249 |
|
BC017355.2 |
- |
3, 4 |
234 |
5020312 |
BC024202.2 |
- |
3, 4 |
234 |
3531069 |
BC035090.2 |
- |
3, 4 |
626 |
5261465 |
D89618.1 |
- |
3, 4 |
301 |
|
DN991804.1 |
- |
3, 4 |
413 |
|
Y12394.1 |
- |
3, 4 |
225 |
|
View Full Product Details