Gene Transcripts

Gene Symbol
RAB9A
Entrez Gene ID
9367
Gene Name
RAB9A, member RAS oncogene family
Gene Aliases
RAB9
Chromosome Location
Chr. X: 13689121 - 13709825 on Build GRCh38
UniGene ID
Hs.495704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195328.1 NP_001182257.1 2 730
NM_004251.4 NP_004242.1 3 819
XM_017029964.1 XP_016885453.1 3 1221
GenBank mRNA AF498944.1 - 1 536
AK290505.1 - 2 709
BC017265.2 - 3 718 4139714
CR450362.1 - 1 536
U44103.1 - 1 536

View Full Product Details