Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000567.2 | NP_000558.2 | 2 | 239 | |
XM_011509207.2 | XP_011507509.1 | 2 | 273 | ||
GenBank mRNA | AK289443.1 | - | 2 | 239 | |
AK313624.1 | - | 2 | 239 | ||
BC020766.1 | - | 2 | 239 | ||
BC125135.1 | - | 2 | 167 | 40119392 | |
BG533281.1 | - | 240 | |||
X56214.1 | - | 2 | 224 | ||
X56692.1 | - | 2 | 224 |