Gene Transcripts

Gene Symbol
FABP7
Entrez Gene ID
2173
Gene Name
fatty acid binding protein 7
Gene Aliases
B-FABP, BLBP, FABPB, MRG
Chromosome Location
Chr. 6: 122749201 - 122784077 on Build GRCh38
UniGene ID
Hs.26770
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319041.1 NP_001305970.1 2 3977
NM_001319042.1 NP_001305971.1 4 567
NM_001446.4 NP_001437.1 4 826
GenBank mRNA AB208815.1 - 2 3724
AJ002962.1 - 4 582
AK091664.1 - 1 2715
BC012299.1 - 4 692 4707233
D50373.1 - 4 558
D88648.1 - 4 557
U51338.1 - 4 561
U81235.1 - 4 521

View Full Product Details