Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001850.4 | NP_001841.2 | 4 | 574 | |
NM_020351.3 | NP_065084.2 | 3 | 512 | ||
GenBank mRNA | AY550974.1 | - | 2 | 372 | |
BC013581.1 | - | 4 | 552 | 3875911 | |
BT009917.1 | - | 1 | 315 | ||
CA393523.1 | - | 3 | 498 | ||
CN484310.1 | - | 3 | 498 |