Gene Transcripts

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
Chromosome Location
Chr. X: 101219786 - 101264497 on Build GRCh38
UniGene ID
Hs.159291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171184.1 NP_001164655.1 22 3109
NM_001939.2 NP_001930.2 24 3392
XM_017029333.1 XP_016884822.1 23 3475
XM_017029334.1 XP_016884823.1 23 3471
GenBank mRNA AB209305.1 - 15 1989
AK289825.1 - 24 3280
U43519.1 - 24 3341

View Full Product Details