Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136109.1 | NP_001129581.1 | 8 | 1119 | |
NM_001136110.1 | NP_001129582.1 | 7 | 867 | ||
NM_001136112.1 | NP_001129584.1 | 9 | 1332 | ||
NM_004347.3 | NP_004338.3 | 9 | 1293 | ||
NR_024239.1 | 7 | 1009 | |||
NR_036562.1 | 6 | 757 | |||
GenBank mRNA | AK296660.1 | - | 8 | 1118 | |
BC074994.2 | - | 8 | 1238 | 30915395 | |
BC113406.1 | - | 8 | 1238 | 8322458 | |
DQ228672.1 | - | 8 | 1261 | ||
DQ228673.1 | - | 7 | 1087 | ||
DQ228674.1 | - | 6 | 835 | ||
DQ228677.1 | - | 8 | 1300 | ||
U28015.1 | - | 8 | 1247 | ||
X94993.1 | - | 8 | 1154 |